SYNM111004
Synthware™ Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 4, Hose Connections: Front-left-right, Rear-left-right
Synonym(s):
Synthware M111004, vacuum manifold, vacuum unit
All Photos(1)
About This Item
Recommended Products
feature
number of ports 4
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M111004
Related Categories
General description
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.