Key Documents
SYNF621460
Synthware pressure equalizing addition funnel
60 mL, joint: ST/NS 14/20, stopcock: PTFE
Synonym(s):
Synthware F621460, graduated addition funnel
Select a Size
1 EA
$281.00
$281.00
Check Cart for Availability
Select a Size
Change View
1 EA
$281.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$281.00
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F621460
capacity
60 mL
60 mL
Related Categories
General description
With 1:5 PTFE stopcock
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)