Skip to Content
Merck
All Photos(1)

Key Documents

SYNF291275

Synthware solvent addition funnel

top diam. 75 mm

Synonym(s):

Synthware F291275, glass funnel

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$92.40

$92.40


Check Cart for Availability


Select a Size

Change View
1 EA
$92.40

About This Item

UNSPSC Code:
41121809
NACRES:
NB.51

$92.40


Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F291275

top diam.

75 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel/packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 90mm in diameter with the stem size as listed.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service