Key Documents
SYNF100003A
Synthware 47 mm filtration apparatus with stainless steel support
Synonym(s):
Synthware 47 mm filtration apparatus with stainless steel support, complete
Select a Size
1 EA
$260.00
$260.00
Check Cart for Availability
Select a Size
Change View
1 EA
$260.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$260.00
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F100003A
Related Categories
General description
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has removable 47 mm stainless steel support screen for use with viscous or proteinaceous solutions. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)