Skip to Content
Merck
All Photos(1)

Key Documents

SM60501010

Sigma-Aldrich

SmartPor, alumina membrane

thickness 50 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$825.00

$825.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
$825.00

About This Item

Linear Formula:
Al
UNSPSC Code:
12352103
NACRES:
NA.23

$825.00


Check Cart for Availability

Request a Bulk Order

material

alumina membrane

description

Pore geometry: trigonal arrangement

packaging

pkg of 10 pieces

size

10 mm × 10 mm

thickness

50 μm

pore size

125 nm (interpore distance)
60 nm pore diameter

compatibility

(Pore geometry: trigonal arrangement)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service