Skip to Content
Merck
All Photos(1)

Key Documents

SM25501010

Sigma-Aldrich

SmartPor, alumina membrane

thickness 50 μm, pore diameter 25 nm, pore size 65 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$769.00

$769.00


Available to ship on29 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$769.00

About This Item

Linear Formula:
Al
UNSPSC Code:
12352103
NACRES:
NA.23

$769.00


Available to ship on29 April 2025Details


Request a Bulk Order

material

alumina membrane

description

Pore geometry: trigonal arrangement

packaging

pkg of 10 pieces

size

10 mm × 10 mm

thickness

50 μm

pore size

25 nm pore diameter
65 nm (interpore distance)

compatibility

(Pore geometry: trigonal arrangement)

Looking for similar products? Visit Product Comparison Guide

Application

Nano porous anodic alumina is a widely studied material used for corrosion protection of aluminum surfaces or as dielectric material in microelectronics applications. Our nano porous anodic alumina exhibits a homogeneous morphology of parallel pores which grow perpendicular to the surface with a narrow distribution of diameters and interpore spacings and possess a standard deviation of <10%. These nano porous alumina find applications, as filters, platforms for multi-functional flow-through sensors, and as templates for the fabrication of nanotubes or nanowires.

Legal Information

Product of SmartMembranes-GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is the alumina membrane(SM25501010) double sided or single sided?

    1 answer
    1. This alumina membrane is double-sided.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service