SLW4510/02
Pyrex® Quickfit(R) Polypropylene screwthread Connector
13 mm Screwthread Inlet and Outlet
Synonym(s):
Pyrex, SciLabware 4510/02
All Photos(3)
About This Item
Recommended Products
material
Viton® O-ring
polypropylene
manufacturer/tradename
4510/02
SciLabware
General description
These screwthread hose connectors are only for use with the QUICKFIT® and PYREX® glassware products. They do not fit DURAN® glass threads.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
PYREX is a registered trademark of Corning, Inc.
QUICKFIT is a registered trademark of DWK Life Sciences
Viton is a registered trademark of E. I. du Pont de Nemours and Company