Skip to Content
Merck
All Photos(1)

Documents

QBD11385

Sigma-Aldrich

Carboxyfluorescein-dPEG ®24-amido-dPEG ®24-DSPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C164H294N3O64P
Molecular Weight:
3363.04
UNSPSC Code:
51171641
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reagent type: fluorescent-labelling reagent

functional group

phospholipid

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

Carboxyfluorescein-dPEG ®₂₄-amido-dPEG ®₂₄-DSPE is a micelle building block with a hydrophilic dPEG spacer attached to a carboxyfluorescein fluorescent dye.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service