P42401
3-Methylpyridine N-oxide
98%
Synonym(s):
3-Methylpyridine 1-oxide, 3-Picoline N-oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
106327
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
form
solid
bp
150 °C/15 mmHg (lit.)
mp
37-39 °C (lit.)
SMILES string
Cc1ccc[n+]([O-])c1
InChI
1S/C6H7NO/c1-6-3-2-4-7(8)5-6/h2-5H,1H3
InChI key
DMGGLIWGZFZLIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service