Skip to Content
Merck
All Photos(2)

Key Documents

183490

Sigma-Aldrich

4-Phenylpyridine N-oxide

98%

Synonym(s):

1-Oxido-4-phenylpyridin-1-ium, 4-Phenylpyridine 1-oxide, 4-Phenylpyridine oxide, p-Phenylpyridine N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$127.00
25 G
$438.00

$127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$127.00
25 G
$438.00

About This Item

Empirical Formula (Hill Notation):
C11H9NO
CAS Number:
Molecular Weight:
171.20
Beilstein:
121493
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

153-155 °C (lit.)

functional group

phenyl

SMILES string

[O-][n+]1ccc(cc1)-c2ccccc2

InChI

1S/C11H9NO/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

VZOPVKZLLGMDDG-UHFFFAOYSA-N

Application

4-Phenylpyridine N-oxide may be used in chemical synthesis studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal Liberka et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
The combinations of Co(II), octacyanidotungstate(V), and monodentate pyridine N-oxide (pyNO) or 4-phenylpyridine N-oxide (4-phpyNO) led to crystallization of novel crystalline phases {CoII[CoII8(pyNO)12(MeOH)12][WV(CN)8]6} (1) and {CoII[CoII8(4-phpyNO)7(MeOH)17][WV(CN)8]6}·7MeOH·(4-phpyNO)3 (2). In both architectures, metal-cyanide clusters are coordinated by N-oxide ligands in a simple monodentate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service