Skip to Content
Merck
All Photos(1)

Key Documents

NCXGSGN660

Sigma-Aldrich

Gold Nanoshells

λmax, 660 nm, NanoXact, 0.05 mg/mL in water, mPEG 5 kDa (Polymer)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$475.00
500 ML
$1,680.00

$475.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$475.00
500 ML
$1,680.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$475.00


Check Cart for Availability

Request a Bulk Order

form

nanoshells

concentration

0.05 mg/mL in water

λmax

660 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our gold nanoshells are surface plasmon resonant (SPR) nanoparticles consisting of a nanoscale silica core surrounded by an ultra-thin gold shell. By changing the ratio of the core diameter and the shell thickness the absorption and scattering properties of the nanoshells can be tuned across the visible and near-IR (NIR) regions. For these shells the increase in the size of the silica core, and decrease in the thickness of the gold shell causes the plasmon resonance to shift toward the NIR region. This class of plasmonic nanoparticles has a wide variety of applications, including uses in optical filters, sensing and cancer therapy. To increase the steric stability our gold nanoshells are coated with PVP (40 kD polyvinylpyrrolidone).

Storage and Stability

DO NOT FREEZE. Store at 2-8C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service