CLS3752
Corning® Thermowell GOLD PCR 96 well plates
96 well plate, Thermowell PCR GOLD plate, full skirt, polypropylene, conical bottom, clear, non-sterile, 50/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
Select a Size
$380.00
Select a Size
About This Item
conical bottom clear wells
skirt
plate format: 96 well, full skirt
$380.00
Recommended Products
material
clear polypropylene plate
conical bottom clear wells
sterility
non-sterile
feature
lid: no
skirt
plate format: 96 well, full skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3752
technique(s)
PCR: suitable
size
96 wells
well volume
200 μL
color
clear
suitability
suitable for (PCR, DNA sequencing and RT-PCR)
application(s)
agriculture
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- 96 well PCR microplates with elevated skirts feature a rigid construction that is ideal for automation
- Clear medical grade polypropylene is suitable for RT-PCR
- Suitable for PCR, DNA sequencing and RT-PCR
- Uniform wall thinness assures efficient heat transfer while minimizing well-to-well variability
- 100% tested for well integrity and certified DNase- and RNase-free
- Nonsterile, without lids
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service