Skip to Content
Merck
All Photos(1)

Key Documents

N9209

Sigma-Aldrich

2′-Nitroacetophenone

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$42.00
25 G
$99.60

$42.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$42.00
25 G
$99.60

About This Item

Linear Formula:
O2NC6H4COCH3
CAS Number:
Molecular Weight:
165.15
Beilstein:
1102322
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$42.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.55 (lit.)

bp

159 °C/16 mmHg (lit.)

mp

23-27 °C (lit.)

SMILES string

CC(=O)c1ccccc1[N+]([O-])=O

InChI

1S/C8H7NO3/c1-6(10)7-4-2-3-5-8(7)9(11)12/h2-5H,1H3

InChI key

SUGXZLKUDLDTKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service