Skip to Content
Merck
All Photos(3)

Key Documents

139351

Sigma-Aldrich

3′-Aminoacetophenone

97%

Synonym(s):

3-Acetylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$56.80
100 G
$180.00

$56.80


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$56.80
100 G
$180.00

About This Item

Linear Formula:
H2NC6H4COCH3
CAS Number:
Molecular Weight:
135.16
Beilstein:
386009
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$56.80


Available to ship on16 April 2025Details


Request a Bulk Order

Assay

97%

bp

289-290 °C (lit.)

mp

94-98 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cccc(N)c1

InChI

1S/C8H9NO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,9H2,1H3

InChI key

CKQHAYFOPRIUOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3′-Aminoacetophenone acts as bifunctional coupling reagent during the synthesis of pyrimidines[1].

Application

3′-Aminoacetophenone (3-Acetylaniline) was used as reagent during the asymmetric total synthesis of pactamycin[1]. It was used as starting reagent during the synthesis of curcumin mimics with substituted sulfonyl group[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chan Mug Ahn et al.
Bioorganic & medicinal chemistry letters, 19(5), 1481-1483 (2009-01-31)
In order to discover novel small vasodilatory molecules for potential use in the treatment of vascular disease, we tested the vasodilatation effect of two types of synthetic curcumin mimics, amide type (3) and sulfonyl amide type (4), upon the basilar
M K Subramanian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(1), 59-67 (2008-01-08)
The gas phase infrared spectrum of 3-aminoacetophenone (3AAP) was measured in the range 5000-500 cm(-1) and with a resolution of 0.5 cm(-1). The Fourier transform Raman (FT-Raman) and Fourier transform infrared (FT-IR) spectra of 3AAP were recorded in the solid
J S Früchtel et al.
Biotechnology and bioengineering, 71(2), 94-103 (2001-04-05)
A combination of symmetric building blocks and combinatorial functional group transformation for synthesis of pyrimidines was investigated. The purpose of the study was to maximize the return on invested synthetic efforts of reaction development for libraries. A representative set of
Justin T Malinowski et al.
Science (New York, N.Y.), 340(6129), 180-182 (2013-04-13)
Medicinal application of many complex natural products is precluded by the impracticality of their chemical synthesis. Pactamycin, the most structurally intricate aminocyclopentitol antibiotic, displays potent antiproliferative properties across multiple phylogenetic domains, but it is highly cytotoxic. A limited number of

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service