All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
180-182 °C (lit.)
SMILES string
[O-][N+](=O)c1ccc2cn[nH]c2c1
InChI
1S/C7H5N3O2/c11-10(12)6-2-1-5-4-8-9-7(5)3-6/h1-4H,(H,8,9)
InChI key
ORZRMRUXSPNQQL-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J X Hao et al.
Pain, 66(2-3), 313-319 (1996-08-01)
We have previously reported that we have observed chronic pain-like response to light mechanical stimuli (allodynia) in rats after severe spinal cord ischemia, which resembles some painful conditions in chronic spinally injured patients and is not relieved by a number
Nobuko Matsumura et al.
The Journal of pharmacology and experimental therapeutics, 325(2), 357-362 (2008-02-14)
7-Nitroindazole (NI) is a widely used inhibitor of neuronal nitricoxide synthase (nNOS) used to study the role of the neuronal NO pathway in the nervous system. 7-NI prevents convulsions, including 2-amino-4-methylphosphinobutyric acid (glufosinate)-induced convulsions, in experimental models. Herein, we examined
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service