Skip to Content
Merck
All Photos(1)

Key Documents

702455

Sigma-Aldrich

1H-Indazole-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$73.20

$73.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$73.20

About This Item

Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$73.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

318-322 °C

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc2[nH]ncc2c1

InChI

1S/C8H6N2O2/c11-8(12)5-1-2-7-6(3-5)4-9-10-7/h1-4H,(H,9,10)(H,11,12)

InChI key

MAVGBUDLHOOROM-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Grailhe et al.
FEBS open bio, 10(10), 2010-2020 (2020-08-19)
Sphingosine-1 phosphate receptor-1 (S1P1 ) activation maintains endothelial barrier integrity, whereas S1P1 desensitization induces peripheral blood lymphopenia. The latter is exploited in the approval and/or late-stage development of receptor-desensitizing agents targeting the S1P1 receptor in multiple sclerosis, such as siponimod
Nikolay T Tzvetkov et al.
European journal of medicinal chemistry, 127, 470-492 (2017-01-21)
The pharmacological and physicochemical analysis of structurally optimized N-alkyl-substituted indazole-5-carboxamides, developed as potential drug and radioligand candidates for the treatment and diagnosis of Parkinson's disease (PD) and other neurological disorders, is reported. Recent efforts have been focused on the development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service