Skip to Content
Merck
All Photos(1)

Key Documents

N14204

Sigma-Aldrich

4-(4-Nitrobenzyl)pyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O2
CAS Number:
Molecular Weight:
214.22
Beilstein:
170877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

69-71 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cc2ccncc2)cc1

InChI

1S/C12H10N2O2/c15-14(16)12-3-1-10(2-4-12)9-11-5-7-13-8-6-11/h1-8H,9H2

InChI key

MNHKUCBXXMFQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in tests for alkylating agents[1] and in a spectroscopic method for phosgene determination.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hemminki et al.
Chemico-biological interactions, 93(1), 51-58 (1994-10-01)
1,2-Epoxy alkanes from C3 to C8 were reacted with DNA, deoxyguanosine and 4-(p-nitrobenzyl) pyridine (NBP). DNA was hydrolyzed at neutral pH to release 7-alkylguanines. The products were analyzed by HPLC. The epoxides reacted largely according to the chain length, shorter
K Futagami et al.
Journal of chromatography. B, Biomedical sciences and applications, 704(1-2), 369-373 (1998-03-28)
We developed a rapid and simple method for identifying 25 commonly used organophosphorus insecticides in human serum using high-performance thin-layer chromatography (HPTLC). These organophosphates were separated on plates with three different developing systems within 6-18 min and detected by means
Shino Manabe et al.
Journal of the American Chemical Society, 124(43), 12638-12639 (2002-10-24)
On-resin real-time monitoring by a combination of a color test of (p-nitrobenzyl)pyridine and Disperse Red was developed for oligosaccharide synthesis.
H B Jiang et al.
Biochemical pharmacology, 60(4), 571-579 (2000-06-30)
Mitomycin C (MC) requires bioreduction prior to the generation of alkylating moieties. NADPH-cytochrome P450 reductase is predominant in metabolic activation of MC in hypoxic cancer cells. In this study, neuronal nitric oxide synthase (nNOS), whose reductase domain is structurally similar
Marina González-Pérez et al.
Chemical research in toxicology, 25(12), 2755-2762 (2012-11-23)
The chemical reactivity of the mutagenic epoxides (EP) propylene oxide (PO), 1,2-epoxybutane (1,2-EB), and cis- and trans-2,3-epoxybutane (cis- and trans-2,3-EB) with 4-(p-nitrobenzyl)pyridine (NBP), a bionucleophile model for S(N)2 alkylating agents with high affinity for the guanine-N7 position, was investigated kinetically.

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service