Skip to Content
Merck
All Photos(2)

Key Documents

8.20890

Sigma-Aldrich

4-(4-Nitrobenzyl)pyridine

for synthesis

Synonym(s):

4-(4-Nitrobenzyl)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$74.40
25 G
$175.00
100 G
$593.00

$74.40


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$74.40
25 G
$175.00
100 G
$593.00

About This Item

Linear Formula:
4-[4-(NO2)C6H4CH2]C5H4N
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12352102
EC Index Number:
214-108-2
NACRES:
NA.22

$74.40


Estimated to ship on08 April 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

crystals

mp

68-71 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)Cc2ccncc2

InChI

1S/C12H10N2O2/c15-14(16)12-3-1-10(2-4-12)9-11-5-7-13-8-6-11/h1-8H,9H2

InChI key

MNHKUCBXXMFQDM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 72 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service