Skip to Content
Merck
All Photos(1)

Key Documents

M78208

Sigma-Aldrich

6-Methylpyridine-2-carboxaldehyde

98%

Synonym(s):

2-Picoline-6-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$350.00

$350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$350.00

About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
107476
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$350.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

refractive index

n20/D 1.527 (lit.)

mp

31-33 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cccc(C=O)n1

InChI

1S/C7H7NO/c1-6-3-2-4-7(5-9)8-6/h2-5H,1H3

InChI key

AHISYUZBWDSPQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muniyandi Sankaralingam et al.
Dalton transactions (Cambridge, England : 2003), 46(22), 7181-7193 (2017-04-19)
A series of Ni(ii) complexes of the types [Ni(L)(CH
Sakthi Raje et al.
Chemistry, an Asian journal, 13(11), 1458-1466 (2018-04-01)
Koneramines (LR OR', R=Ph or Ts; R'=Me, iPr) and their complexes were found to emerge from the system of pyridine-2-carboxaldehyde and N-phenyl/tosylethylenediamine when a primary or secondary alcohol was used as solvent. Imidazolidinylpyridines (LR , R=Ph or Ts) became major

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service