Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Novel nickel(ii) complexes of sterically modified linear N4 ligands: effect of ligand stereoelectronic factors and solvent of coordination on nickel(ii) spin-state and catalytic alkane hydroxylation.
Novel nickel(ii) complexes of sterically modified linear N4 ligands: effect of ligand stereoelectronic factors and solvent of coordination on nickel(ii) spin-state and catalytic alkane hydroxylation.
Dalton transactions (Cambridge, England : 2003) (2017-04-19)
Muniyandi Sankaralingam, Prabha Vadivelu, Mallayan Palaniandavar
ABSTRACT
A series of Ni(ii) complexes of the types [Ni(L)(CH
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%
Sigma-Aldrich
6-Methylpyridine-2-carboxaldehyde, 98%
Sigma-Aldrich
1-Methyl-2-imidazolecarboxaldehyde, 98%