Skip to Content
Merck
All Photos(1)

Key Documents

M153

Sigma-Aldrich

Maleic acid

99%

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

130-135 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Wei Wei et al.
Journal of colloid and interface science, 505, 392-401 (2017-06-18)
Synthetic sugar alcohol mono-behenates with high melting points, surface activity and resistance to enzymatic lipolysis, are expected to form stable single-component solid lipid nanocarriers (SC-SLNs). The preparation methods and the polar head group of the molecules should affect the size
Cui Li et al.
Journal of pharmaceutical and biomedical analysis, 163, 105-112 (2018-10-07)
A new ultra-high performance liquid chromatography combined with triple quadrupole mass spectrometry was developed to evaluate the quality of Tanreqing injection. Seven flavonoids (Rutin, Baicalin, Scutellarin, Chrysin-7-O-Beta-d-glucoronide, Oroxylin A-7-O-β-d-glucoronide, Wogonin, Luteolin-7-O-glucoside), two phenolic acids (Chlorogenic acid, Caffeic acid) and two
K Grundke et al.
Advances in colloid and interface science, 222, 350-376 (2014-12-10)
Contact angle hysteresis phenomena on polymer surfaces have been studied by contact angle measurements using sessile liquid droplets and captive air bubbles in conjunction with a drop shape method known as Axisymmetric Drop Shape Analysis - Profile (ADSA-P). In addition
Hanna Kotikoski et al.
Acta ophthalmologica Scandinavica, 81(4), 355-360 (2003-07-16)
To investigate whether a nitric oxide donor given as a single oral dose is able to modify aqueous humour flow in healthy volunteers. Ten healthy volunteers participated in a randomized, double-masked and placebo-controlled cross-over study. Aqueous humour flow was assessed
J M Herreras et al.
Ophthalmology, 99(7), 1082-1088 (1992-07-01)
This study was undertaken to see whether long-term locally applied ocular medications produced any alterations in the ocular surface, and, in particular, whether it caused damage to the mucus layer of the tear film. The authors studied the ocular surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service