Skip to Content
Merck
All Photos(2)

Documents

8.00380

Sigma-Aldrich

Maleic acid

for synthesis

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-742-5
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99% (acidimetric)

form

powder

potency

708 mg/kg LD50, oral (Rat)
1560 mg/kg LD50, skin (Rabbit)

pH

1.3 (20 °C, 100 g/L in H2O)

mp

130-135 °C (lit.)

solubility

478.8 g/L

density

1.59 g/mL at 25 °C (lit.)

bulk density

750‑800 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Food industry metabolomics: Maleic acid is identified as a key metabolite in the profiling of jelly fungi, aiding in the determination of biomarkers for different flesh colors, which is vital for quality assessment in the food industry (Ye et al., 2024).
  • Chromatography device innovation: Maleic acid contributes to the design and characterization of an electrochemically-modulated membrane chromatography device, enhancing the precision and functionality of separation technologies in analytical chemistry (Röcker et al., 2024).


Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 130 °C
Melting range (upper value): ≤ 135 °C
Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service