Skip to Content
Merck
All Photos(1)

Documents

HAM238100

Hamilton® MiniTrode lab pH electrode

glass, S7 connector

Synonym(s):

Hamilton pH Electrode, Mini pH Electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.79

material

ceramic diaphragm
cylindrical HF glass membrane
glass

feature

autoclavable: no
electrode 3 M KCl electrolyte
S7 connector

parameter

0-100 °C temp. range

diam.

12 mm

immersion depth

7 mm

pH

0-14

General description

  • Hamilton® MiniTrode lab pH electrode is optimized for pH measurements in very small volumes (e.g., vials).
  • Requires an immersion depth of only 7 mm.
  • Long-term stable Everef system.

Application

Hamilton® MiniTrode lab pH electrode has been used to measure the pH of:

  • plasma-treated water
  • diluted bull semen and SP4medium
  • Saccharomyces cerevisiae strain suspensions

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Addition of Lactobacillus spp., Enrofloxacin or Doxycycline Negatively Affects the Viability of Mycoplasma bovis in Diluted Bovine Semen
Ana Garcia G, et al.
Animals : an open access journal from MDPI, 10(5) (2020)
Evolutionary clade affects resistance of Clostridium difficile spores to Cold Atmospheric Plasma
Mairead C, et al.
Scientific Reports (2017)
Jakub Suchodolski et al.
Microorganisms, 7(10) (2019-09-25)
Candida albicans is an opportunistic fungal pathogen of humans. Treatment of C. albicans infections relies on azoles, which target the lanosterol 14α-demethylase (Erg11p) encoded by the ERG11 gene. Our results show that targeted gene disruption of ERG11 can result in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service