Z113255
Sigma-Aldrich® standard Ag/AgCl double junction pH combination electrode, glass body
Standard pin, electrode L 55 mm , with bulb guard
Select a Size
$363.00
Select a Size
About This Item
$363.00
Recommended Products
electrode L
55 mm , with bulb guard
stem O.D.
8 mm
General description
Temperature Range:
- -5 to 100°C Continuous Use
- -5 to 130°C Intermittent Use
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Features and Benefits
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)