Skip to Content
Merck
All Photos(1)

Key Documents

DWKWCL1000-3

WHEATON® CELLine Multiuse Bioreactor flasks for suspension cell culture

polystyrene, flask capacity (1,000 mL), case of 3 ea

Synonym(s):

CELLine Bioreactor

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$1,710.00

$1,710.00


Available to ship on29 April 2025Details



Select a Size

Change View
3 EA
$1,710.00

About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

$1,710.00


Available to ship on29 April 2025Details


material

molded polypropylene neck
polystyrene
screw top plastic cap

sterility

sterile; γ-irradiated

feature

with cap

packaging

case of 3 ea

manufacturer/tradename

WHEATON® WCL1000-3

flask capacity

(1,000 mL)

Looking for similar products? Visit Product Comparison Guide

General description

CELLine Bioreactor Flask for high density suspension cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines

  • Disposable and ready-to-use
  • High call density and high product concentration
  • Bulk media chamber allows for longer run times without media refreshing and reduces operation time
  • Decreases use of consumables
  • Cost-efficient, space saving, and stackable
  • No additional equipment required for operation

Features and Benefits

  • Ideal for high density cell culture, designed to enhance small scale bio-production for antibody and protein generation_x000D_
  • Available in 1000ml flask for suspension (CELLine 1000) or adherent (CELLine 1000-AD) cell culture and 350ml flask for suspension culture only_x000D_
  • Disposable and ready-to-use_x000D_
  • Multi-harvest operation ensures a cost efficient solution for antibody and protein production_x000D_
  • Ideal for use with cell lines such as Hybridoma, CHO, HEK, BHK, Sf9

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service