Skip to Content
Merck
All Photos(3)

Key Documents

DWKFDF112A

Azlon Clear PMP Volumetric Flask with PPE Stopper

class A, tolerance, capacity 25 mL

Synonym(s):

FDF112A

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$75.50

$75.50


Check Cart for Availability


Select a Size

Change View
1 EA
$75.50

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

$75.50


Check Cart for Availability

material

round clear PMP flask

feature

class A tolerance

packaging

pkg of 1 ea

manufacturer/tradename

Azlon FDF112A

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

Application

For the accurate preparation of solutions where glass alternatives would not be suitable

Features and Benefits

Excellent clarity and calibrated to ISO 1042

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service