Skip to Content
Merck
All Photos(1)

Key Documents

DWKBLP0125PGRA

Azlon Polypropylene Graduated Wide Neck Round Bottle with Write-On Panel

screw cap, round translucent polypropylene bottle, capacity 125 mL

Synonym(s):

BLP0125PGRAD

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$70.00

$70.00


Check Cart for Availability


Select a Size

Change View
10 EA
$70.00

About This Item

UNSPSC Code:
41121818
NACRES:
NB.43

$70.00


Check Cart for Availability

material

round translucent polypropylene bottle
screw cap
wide-neck

sterility

non-sterile

packaging

pkg of 10 ea

manufacturer/tradename

Azlon BLP0125PGRA

capacity

125 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for management and storage of both liquid and powder samples

Features and Benefits

Graduated write on panel

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service