Skip to Content
Merck
All Photos(1)

Key Documents

D98807

Sigma-Aldrich

N,N-Diethylnicotinamide

99%

Synonym(s):

DENC, Nicotinic acid diethylamide, ‘Nikethamide’

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$58.90
500 G
$260.00

$58.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$58.90
500 G
$260.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2O
CAS Number:
Molecular Weight:
178.23
Beilstein:
5743
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.90


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.524 (lit.)

bp

296-300 °C (lit.)

mp

23 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(=O)c1cccnc1

InChI

1S/C10H14N2O/c1-3-12(4-2)10(13)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3

InChI key

NCYVXEGFNDZQCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diethylnicotinamide (DENC) is commonly used as a ligand to prepare tetranuclear copper complexes.[1][2][3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of mixed-valence copper molecules (. mu.-Y) N4CuII2CUI2X4 (N= N, N-diethylnicotinamide; Y= 3, 4, 5, 6-tetrachlorocatecholate; X= Cl, Br, I) and the products and kinetics of their reactions with dioxygen and nitrobenzene.
El-Sayed M A and Davies G
Inorganic Chemistry, 29(24), 4891-4897 (1990)
Transmetalation reactions of tetranuclear copper (II) complexes. 2. Stoichiometry and products of reaction of [(DENC) CuCl] 4O2,[(DENC) CuCl] 4 (CO3) 2,[(DENC) CuCl] 4Cl4, and (DENC) 4Cu4Cl6O (DENC= N, N-diethylnicotinamide) with Ni (NS) 2, the kinetics of product isomerization in aprotic solvents, and inhibition of copper-catalyzed phenolic. oxidativecoupling by dioxygen through transmetalation.
El-Toukhy A, et al.
Journal of the American Chemical Society, 106(16), 4596-4605 (1984)
Synthesis, structure and properties of the tetranuclear complexes [(DENC) CuX] 4 (DENC= N, N-diethylnicotinamide; X= Cl, Br, I) and the kinetics of oxidation of the chloride and bromide by dioxygen in aprotic solvents.
Churchill M R, et al.
Inorganic Chemistry, 21(3), 995-1001 (1982)
Tooru Ooya et al.
Journal of controlled release : official journal of the Controlled Release Society, 93(2), 121-127 (2003-11-26)
New methods and pharmaceutical compositions were developed to increase the aqueous solubility of paclitaxel (PTX), a poorly water-soluble drug. Graft and star-shaped graft polymers consisting of poly(ethylene glycol) (PEG400) graft chains increased the PTX solubility in water by three orders
X H Ren et al.
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases, 16(6), 331-334 (1993-12-01)
Twelve subjects with COPD in acute exacerbation of respiratory failure were studied. The experiment of each subject was divided into three steps: room air breathing, 35% O2 inhalation for one hour, and then intravenous drip of nikethamide (1.875g) for two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service