Skip to Content
Merck
All Photos(1)

Documents

367095

Sigma-Aldrich

4-(Ethylaminomethyl)pyridine

97%

Synonym(s):

N-(Pyridin-4-ylmethyl)ethanamine, N-Ethyl-4-pyridinemethanamine, NSC 85903

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.516 (lit.)

density

0.979 g/mL at 25 °C (lit.)

SMILES string

CCNCc1ccncc1

InChI

1S/C8H12N2/c1-2-9-7-8-3-5-10-6-4-8/h3-6,9H,2,7H2,1H3

InChI key

ZBAMQLFFVBPAOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Ethylaminomethyl)pyridine is a secondary amine.

Application

4-(Ethylaminomethyl)pyridine is suitable for use in the synthesis of sodium dithiocarbamate ligands. It may be used in the synthesis of 4-(ethylaminodithiocarbamate) methylpyridine diorganotin derivatives. It may be used in the synthesis of {[ethyl(pyridin-4-ylmethyl)carbamothioyl]sulfanido-κ2 S,S′}(1,4,7,10,13,16-hexaoxacyclooctadecane-κ6O)potassium.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adduct effects on structure and luminescence in a series of new dithiocarbamatogold (I) complexes. T
Han S, et al.
Transition Met. Chem. (London), 36(7), 691-697 (2011)
Synthesis and structural characterization of diorganotin dithiocarbamates from 4-(ethylaminomethyl) pyridine.
Barba V, et al.
Heteroatom Chem., 23(5), 422-428 (2012)
Hadi D Arman et al.
Acta crystallographica. Section E, Structure reports online, 69(Pt 9), m479-m480 (2014-01-16)
The asymmetric unit of title salt co-crystal, [K(C9H11N2S2)(C12H24O6)], comprises a K(+) cation, an (-)S2CN(Et)py anion and a 18-crown-6 mol-ecule. Substantial delocalization of π-electron density is evident in the di-thio-carbamate anion, as indicated by the equivalent C-S bond lengths. The K(+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service