Skip to Content
Merck
All Photos(2)

Key Documents

D91683

Sigma-Aldrich

O,O′-Diethyl chlorothiophosphate

97%

Synonym(s):

O,O′-Diethylthiophosphoric chloride, Diethyl phosphorochloridothionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$84.40

$84.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$84.40

About This Item

Linear Formula:
(CH3CH2O)2P=(S)Cl
CAS Number:
Molecular Weight:
188.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.40


Check Cart for Availability

Request a Bulk Order

vapor pressure

1.4 mmHg ( 50 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

45 °C/3 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

CCOP(Cl)(=S)OCC

InChI

1S/C4H10ClO2PS/c1-3-6-8(5,9)7-4-2/h3-4H2,1-2H3

InChI key

KMJJJTCKNZYTEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kataoka et al.
Journal of chromatography. A, 723(1), 93-99 (1996-02-02)
A selective and sensitive method was developed for the determination of volatile N-nitrosamines by gas chromatography (GC). After denitrosation of N-nitrosamines with hydrobromic acid, the resulting secondary amines were converted into their N-diethylthiophosphoryl derivatives and then measured by GC using
Mateusz Daśko et al.
Drug development research, 80(6), 857-866 (2019-07-14)
In the present work, we described convenient methods for the synthesis of N-thiophosphorylated 3-(4-aminophenyl)-coumarin-7-O-sulfamates as steroid sulfatase (STS) inhibitors. To design the structures of the potential STS inhibitors, molecular modeling techniques were used. A computational docking method was used to
H Kataoka et al.
Biomedical chromatography : BMC, 7(3), 129-133 (1993-05-01)
A selective and sensitive method has been developed for the determination of secondary amines by gas chromatography (GC). After removal of primary amines by the reaction with o-phthaldialdehyde, secondary amines were converted into their N-diethylthiophosphoryl derivatives and then measured by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service