Skip to Content
Merck
All Photos(3)

Key Documents

344249

Sigma-Aldrich

Diphenyl phosphoryl chloride

96%

Synonym(s):

Diphenyl chlorophosphate, Diphenyl phosphorochloridate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$95.30
500 G
$312.00

$95.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$95.30
500 G
$312.00

About This Item

Linear Formula:
(C6H5O)2POCl
CAS Number:
Molecular Weight:
268.63
Beilstein:
654130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$95.30


Check Cart for Availability

Request a Bulk Order

vapor pressure

10 mmHg ( 190 °C)

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.55 (lit.)

bp

314-316 °C/272 mmHg (lit.)

density

1.296 g/mL at 25 °C (lit.)

SMILES string

ClP(=O)(Oc1ccccc1)Oc2ccccc2

InChI

1S/C12H10ClO3P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H

InChI key

BHIIGRBMZRSDRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyl phosphoryl chloride is a diaryl phosphoryl derivative. It is formed during the preparation of monophenyl phosphoryl chloride via reaction of phenol and phosphorus oxychloride.[1]

Application

Diphenyl phosphoryl chloride may be used:
  • in the preparation of guanosine-adenosine-5′,5′-triphosphate, guanosine-cytidine-5′,5′-triphosphate, guanosine-uridine-5′,5′-triphosphate, diguanosine-5′,5′-diphosphate and diguanosine-5′,5′-triphosphate[2]
  • as phosphorylating agent for the synthesis of 2-deoxy-D-galactose-3 and -6 phosphoric acids[3]
  • in the synthesis of diaryl phosphoryl derivatives of alkylene dithiophosphates[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dialkyl (aryl) phosphoryl derivatives of alkylene dithiophosphates.
Sharma CS and Nagar PN.
Phosphorus, Sulfur, and Silicon and the Related Elements, 179(9), 1793-1798 (2004)
219. Deoxy-sugars. Part XV. D-Galactose-3 and-6 phosphoric acids and their 2-deoxy-analogues.
Foster AB, et al.
Journal of the Chemical Society, 980-987 (1951)
Synthesis of Disodium Phenyl Phosphate.
Freeman HF and Colver CW.
Journal of the American Chemical Society, 60(4), 750-751 (1938)
Y Furuichi et al.
The Journal of biological chemistry, 251(16), 5043-5053 (1976-08-25)
Blocked and methylated 5' termini of reovirus mRNA are formed by viral cores at an early stage of transcription. Cores incubated in a complete transcription reaction mixture for 30 s, or in a mixture lacking UTP and ATP for a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service