Skip to Content
Merck
All Photos(1)

Key Documents

D308

Sigma-Aldrich

Decahydro-2-naphthol

97%

Synonym(s):

2-Decalol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$71.60

$71.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$71.60

About This Item

Linear Formula:
C10H17OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
773927
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.60


Check Cart for Availability

Request a Bulk Order

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.5 (lit.)

bp

109 °C/14 mmHg (lit.)

density

0.996 g/mL at 25 °C (lit.)

SMILES string

OC1CCC2CCCCC2C1

InChI

1S/C10H18O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h8-11H,1-7H2

InChI key

UPMAOXLCTXPPAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can you please provide any information on stereochemistry of this chemical? Is it cis- or trans isomer? We have bought it but no details on this topic was provided in the specification sheet.

    1 answer
    1. Unfortunately, the stereochemistry is not determined for this product.

      Helpful?

Reviews

No rating value

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service