Skip to Content
Merck
All Photos(1)

Key Documents

CDS000161

Sigma-Aldrich

4-Aminocyclohexanecarboxylic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$147.00

$147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$147.00

About This Item

Empirical Formula (Hill Notation):
C7H13NO2
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

$147.00


Check Cart for Availability

Request a Bulk Order

form

solid

SMILES string

NC1CCC(CC1)C(O)=O

InChI

1S/C7H13NO2/c8-6-3-1-5(2-4-6)7(9)10/h5-6H,1-4,8H2,(H,9,10)

InChI key

DRNGLYHKYPNTEA-UHFFFAOYSA-N

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Lv et al.
Journal of controlled release : official journal of the Controlled Release Society, 320, 412-420 (2020-02-06)
Cytosolic protein delivery is important for the development of protein therapeutics towards intracellular targets. Guanidyl polymers exhibit high binding affinity with cargo proteins, and thus were designed as carriers for intracellular protein delivery. However, the structure-activity relationship and mechanism of
Nađa Kostić et al.
Journal of pharmaceutical and biomedical analysis, 109, 79-84 (2015-03-15)
In this paper, novel LC-MS/MS methods for the determination of antiepileptic drug pregabalin in dried matrix spots (DMS) are presented. This attractive technique of sample collection in micro amount was utilized in the form of dried blood spots (DBS) and
Nađa Kostić et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 962, 102-108 (2014-06-09)
This paper presents a LC-MS/MS method for the determination of antiepileptic drug vigabatrin in dried plasma spots (DPS). Due to its zwitterionic chemical structure, a pre-column derivatization procedure was performed, aiming to yield enhanced ionization efficiency and improved chromatographic behaviour.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service