Skip to Content
Merck
All Photos(1)

Key Documents

D145203

Sigma-Aldrich

3-Dimethylamino-1-propyl chloride hydrochloride

96%

Synonym(s):

3-Chloro-N,N-dimethylpropylamine hydrochloride, N,N-Dimethyl(3-chloropropyl)amine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$92.50

$92.50


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$92.50

About This Item

Linear Formula:
(CH3)2N(CH2)3Cl · HCl
CAS Number:
Molecular Weight:
158.07
Beilstein:
3670046
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.50


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

96%

form

crystals

mp

140-143 °C (lit.)

SMILES string

Cl[H].CN(C)CCCCl

InChI

1S/C5H12ClN.ClH/c1-7(2)5-3-4-6;/h3-5H2,1-2H3;1H

InChI key

LJQNMDZRCXJETK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Carfagna et al.
Molecular pharmacology, 44(1), 129-141 (1993-07-01)
Perturbations of rat brain synaptic plasma membrane (SPM) bilayer structure and Na+/K(+)-ATPase activity were correlated for drugs that are structurally related and exhibit similar toxicological side effects but belong to different pharmacological classes. Na+/K(+)-ATPase IC50 values decrease linearly with increasing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service