Skip to Content
Merck
All Photos(1)

Key Documents

C47809

Sigma-Aldrich

5-Chloroindole-2-carboxylic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$85.70

$85.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$85.70

About This Item

Empirical Formula (Hill Notation):
C9H6ClNO2
CAS Number:
Molecular Weight:
195.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$85.70


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

287 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc2cc(Cl)ccc2[nH]1

InChI

1S/C9H6ClNO2/c10-6-1-2-7-5(3-6)4-8(11-7)9(12)13/h1-4,11H,(H,12,13)

InChI key

FUQOTYRCMBZFOL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Pralong et al.
Neuroscience letters, 136(1), 56-58 (1992-02-17)
The potency, specificity and reversibility of various presumed glycine site N-methyl-D-aspartate (NMDA) antagonists was studied on neonatal rat spinal cord using the grease gap technique. 5,7-Dichlorokynurenate was the most potent and specific glycine site antagonist among the compounds tested. On

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service