CLS763019
Pyrex® PTFE key-hole stopper, color-coded
green, stopper size 19
Synonym(s):
flask stopper, flask stoppers, stoppers, Corning® 7630
Select a Size
$125.00
Select a Size
About This Item
$125.00
Recommended Products
stopper size
19
manufacturer/tradename
Corning 7630-19
color
green
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service