Key Documents
CLS762419
Pyrex® HDPE stopper, improved form
stopper joint: 19
Synonym(s):
flask stopper, flask stoppers, stoppers, Corning® 7624
Select a Size
6 EA
$23.60
$23.60
Available to ship on28 April 2025Details
Select a Size
Change View
6 EA
$23.60
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
$23.60
Available to ship on28 April 2025Details
Joint
joint: stopper19
manufacturer/tradename
Corning 7624-19
General description
Blue and white stoppers with closed bottoms, made of linear high-density polyethylene. Improved design fully protects the neck in the event of accidental tipping.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)