Key Documents
CLS762438
Pyrex® HDPE stopper, improved form
stopper joint: 38
Synonym(s):
flask stopper, flask stoppers, stoppers, Corning® 7624
Select a Size
6 EA
$41.50
$41.50
Check Cart for Availability
Select a Size
Change View
6 EA
$41.50
About This Item
UNSPSC Code:
41122500
$41.50
Check Cart for Availability
Joint
joint: stopper38
manufacturer/tradename
Corning 7624-38
General description
Blue and white stoppers with closed bottoms, made of linear high-density polyethylene. Improved design fully protects the neck in the event of accidental tipping.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)