CLS70775N
Pyrex® disposable serological pipettes
volume 5.0 mL
Synonym(s):
Corning® 7077
Select a Size
$239.00
Select a Size
About This Item
$239.00
Recommended Products
material
borosilicate glass
sterility
sterile
feature
DNase free: no
RNase free: no
graduations
subdivision
packaging
pack of 120 ea
case of 720 ea
manufacturer/tradename
Corning 7077-5N
measuring range
accuracy: 0.1 mL
capacity
5.0 mL
volume
5.0 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® disposable borosilcate glass serological pipettes are calibrated "to deliver" with blow-out.
- Features long, slender, tapered tips for faster and easier pipetting.
- Includes negative graduations for additional working volume.
- Fire-polished tips for burr-free, even flow rates.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service