Skip to Content
Merck
All Photos(1)

Key Documents

CLS7071010

Pyrex® VISTA volumetric pipette, Class A

volume 10 mL

Synonym(s):

Corning® 70710

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$184.00

$184.00


Available to ship on23 April 2025Details



Select a Size

Change View
12 EA
$184.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

$184.00


Available to ship on23 April 2025Details


material

glass

feature

class A
graduation

packaging

case of 12 ea
pack of 12 ea

manufacturer/tradename

Corning 70710-10

volume measuring range

10 mL

capacity

10 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service