CLS630242L
PYREXPLUS® single metric scale graduated cylinders, with protective coating
volume 2,000 mL
Synonym(s):
Corning® 63024
Select a Size
$485.00
Select a Size
About This Item
$485.00
Recommended Products
feature
subdivision 20.0 mL
manufacturer/tradename
Corning 63024-2L
tol. measuring range
10.0 mL
volume
2,000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Calibrated "to deliver"
- Single metric scale
- Protective plastic coating to help prevent glass from shattering & reduces spills
- Autoclavable (121 °C)
- Resistant to thermal shock
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service