Key Documents
CLS5320500
Pyrex® heavy-wall graduated filtering flask
capacity 500 mL
Synonym(s):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
Select a Size
18 EA
$430.00
$430.00
Check Cart for Availability
Select a Size
Change View
18 EA
$430.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$430.00
Check Cart for Availability
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper7
description
without hose connection
sterility
non-sterile
feature
closure type Rubber stopper
packaging
case of 18
pack of 6
manufacturer/tradename
Corning 5320-500
volume measuring range
150-500 mL
capacity
500 mL
General description
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)