CLS51395500
Pyrex® low actinic media storage bottle, graduated with screw cap
capacity 500 mL
Synonym(s):
graduated bottle, media bottle, media bottles, pyrex bottle, Corning® 51395
Select a Size
$372.00
Estimated to ship on13 April 2025
Select a Size
About This Item
$372.00
Estimated to ship on13 April 2025
Recommended Products
material
amber bottle
borosilicate glass bottle
cylindrical bottle
polypropylene cap
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
graduations
packaging
case of 4 ea
pack of 4 ea
manufacturer/tradename
Corning 51395-500
availability
not available in EU
bottle neck diam.
30 mm
capacity
500 mL
diam. × H
86 mm × 176 mm
fitting
45
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service