CLS304210
Pyrex® Lifetime Red™ single metric scale graduated cylinder
volume 10 mL
Synonym(s):
Corning® 3042
Select a Size
$586.00
Estimated to ship on29 April 2025
Select a Size
About This Item
$586.00
Estimated to ship on29 April 2025
Recommended Products
feature
subdivision 0.1 mL
manufacturer/tradename
Corning 3042-10
tol. measuring range
0.1 mL
volume
10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Calibrated "to contain"
- Single metric scale
- White enamel scale against red band
- Removable PE bumperguard
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service