CLS30262L
Pyrex® class A double metric scale graduated cylinder, with blue enamel
volume 2,000 mL
Synonym(s):
Corning® 3026
Select a Size
$170.00
Select a Size
About This Item
$170.00
Recommended Products
feature
subdivision 20.0 mL
manufacturer/tradename
Corning 3026-2L
tol. measuring range
6.0 mL
volume
2,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service