CLS1010
Pyrex® beaker with recessed wall for handle
complete
Synonym(s):
Corning container
Select a Size
$2,690.00
Select a Size
About This Item
$2,690.00
Recommended Products
material
borosilicate glass
feature
graduations: no
manufacturer/tradename
Corning 1010
capacity
3000 mL
Related Categories
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
What are the dimensions of product CLS1010, 3,000 mL PYREX® Beaker with Recessed Wall for Handle, Complete?
Product CLS1010, 3,000 mL PYREX® Beaker with Recessed Wall for Handle, Complete, is approximately 213 mm tall and 155 mm in diameter.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.