Skip to Content
Merck
All Photos(1)

Documents

CDS019977

Sigma-Aldrich

Aldol

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CC(O)CC=O

InChI

1S/C4H8O2/c1-4(6)2-3-5/h3-4,6H,2H2,1H3

InChI key

HSJKGGMUJITCBW-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Eye Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Immacolata Luisi et al.
PloS one, 8(2), e56469-e56469 (2013-03-02)
We have identified a 101-amino-acid polypeptide derived from the sequence of the IIA binding site of human albumin. The polypeptide contains residues that make contact with IIA ligands in the parent protein, and eight cysteine residues to form disulfide bridges
Christopher P Lakeland et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(1), 155-159 (2019-10-28)
Sydnone-based cycloaddition reactions are a versatile platform for pyrazole synthesis, however they operate under harsh conditions (high temperature and long reaction times). Herein we report a strategy that addresses this limitation utilizing the synergistic combination of organocatalysis and visible-light photocatalysis.
Ian Paterson et al.
Organic & biomolecular chemistry, 13(20), 5716-5733 (2015-04-23)
The brasilinolides are an architecturally complex family of 32-membered macrolides, characterised by potent immunosuppressant and antifungal properties, which represent challenging synthetic targets. By adopting a highly convergent strategy, a range of asymmetric aldol/reduction sequences and catalytic protocols were employed to
Martha A Grover et al.
Life (Basel, Switzerland), 9(3) (2019-07-04)
Living systems employ both covalent chemistry and physical assembly to achieve complex behaviors. The emerging field of systems chemistry, inspired by these biological systems, attempts to construct and analyze systems that are simpler than biology, while still embodying biological design
Naoki Ishida et al.
Angewandte Chemie (International ed. in English), 57(35), 11399-11403 (2018-07-19)
An electronically neutral 2-arylsilacyclobutane generates a nucleophilic carbanion at room temperature through cleavage of the benzylic C-Si bond when simply dissolved in polar aprotic solvents such as N,N-dimethylformamide (DMF). The nucleophilic species is capable of capturing carbon dioxide to furnish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service