Skip to Content
Merck
All Photos(1)

Key Documents

479284

Sigma-Aldrich

(R)-(−)-O-Formylmandeloyl chloride

97%

Synonym(s):

(−)-O-Formyl-D-mandeloyl chloride, (R)-(−)-α-(Formyloxy)phenylacetyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$213.00

$213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$213.00

About This Item

Linear Formula:
HCO2CH(C6H5)COCl
CAS Number:
Molecular Weight:
198.60
Beilstein:
5404068
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$213.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

optical activity

[α]23/D −228°, neat

refractive index

n20/D 1.523 (lit.)

bp

221 °C (lit.)

density

1.275 g/mL at 25 °C (lit.)

functional group

acyl chloride
phenyl

storage temp.

2-8°C

SMILES string

ClC(=O)[C@H](OC=O)c1ccccc1

InChI

1S/C9H7ClO3/c10-9(12)8(13-6-11)7-4-2-1-3-5-7/h1-6,8H/t8-/m1/s1

InChI key

ZNLABNPTWSKGDX-MRVPVSSYSA-N

Application

(R)-(−)-O-Formylmandeloyl chloride can be used as a chiral resolving agent for the resolution of spirobiindane.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and application of a new hexamethyl-1, 1′-spirobiindane-based chiral bisphosphine (HMSI-PHOS) ligand in asymmetric allylic alkylation
Chang S, et al.
Organic & Biomolecular Chemistry, 16(13), 2239-2247 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service