Skip to Content
Merck
All Photos(1)

Key Documents

CDS018221

Sigma-Aldrich

2-(undecyloxy)ethanol

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$276.00

$276.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$276.00

About This Item

Empirical Formula (Hill Notation):
C13H28O2
Molecular Weight:
216.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$276.00


Check Cart for Availability

Request a Bulk Order

form

solid

SMILES string

CCCCCCCCCCCOCCO

InChI

1S/C13H28O2/c1-2-3-4-5-6-7-8-9-10-12-15-13-11-14/h14H,2-13H2,1H3

InChI key

KLMGMPDXSPSCOC-UHFFFAOYSA-N

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph C H Wong et al.
Journal of economic entomology, 110(5), 2269-2274 (2017-09-30)
Here, we describe several field experiments that evaluated potential problems with current methods of trapping cerambycid beetles using panel traps baited with synthesized pheromones. Positioning traps at least 5 m apart in linear transects was effective in preventing unbaited traps
Bridget Bobadoye et al.
Environmental entomology, 48(1), 189-192 (2018-12-06)
During field bioassays in Kenya of 10 chemicals that are common pheromone components of cerambycid beetles, six species in the subfamily Cerambycinae were significantly attracted to 3-hydroxyhexan-2-one, and one species in the subfamily Lamiinae was significantly attracted to 2-(undecyloxy)ethanol (known
J C H Wong et al.
Environmental entomology, 46(3), 649-653 (2017-04-13)
Here, we describe a field experiment that tested for attraction of cerambycid beetles to odors from angiosperm hosts, and whether plant volatiles also serve to enhance attraction of beetles to their aggregation-sex pheromones. Traps were baited with a blend of
Claudia D Lyons-Yerion et al.
Environmental entomology, 49(5), 1077-1087 (2020-09-05)
Research over the last 15 yr has shown widespread pheromone parsimony within the coleopteran family Cerambycidae, with a number of highly conserved pheromone motifs, often shared within and across subfamilies, tribes, and genera. Our goals were to increase our understanding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service