Skip to Content
Merck
All Photos(1)

Key Documents

G401000

Sigma-Aldrich

DMT-dG(ib)-CPG

pore size 1000 Å, 25-35μmol/g

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$55.50
10 G
$496.00

$55.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$55.50
10 G
$496.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

$55.50


Check Cart for Availability

Request a Bulk Order

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service